Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080338.1 | NP_001073807.1 | 1 - 2 | 134 | 65 | |
GenBank mRNA | BC122676.1 | - | 1 - 2 | 134 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |