Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMED10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040549.2 | NP_001035639.1 | 1 - 2 | 254 | 84 | |
GenBank mRNA | BC123529.1 | - | 1 - 2 | 254 | 84 | |
BT021049.1 | - | 1 - 2 | 255 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |