Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082438.2 | NP_001075907.1 | 1 - 2 | 302 | 84 | |
GenBank mRNA | BC133568.1 | - | 1 - 2 | 303 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |