Search
Search

TBCCD1| Gene Symbol: TBCCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083730.1 | NP_001077199.1 | 8 - 9 | 1903 | 114 | |
| GenBank mRNA | BC134464.1 | - | 8 - 9 | 1903 | 114 | |
Set Membership: |
|