Search Thermo Fisher Scientific
Gene Symbol: FRMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098985.2 | NP_001092455.1 | 5 - 6 | 541 | 92 | |
GenBank mRNA | BC142304.1 | - | 5 - 6 | 542 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |