Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTCFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123365.1 | NP_001116837.1 | 2 - 3 | 488 | 70 | |
GenBank mRNA | EU527855.1 | - | 2 - 3 | 488 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |