Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CRAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205874.1 | NP_001192803.1 | 18 - 19 | 3514 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |