Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206024.1 | NP_001192953.1 | 1 - 2 | 78 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |