Search Thermo Fisher Scientific
Gene Symbol: KSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206862.1 | NP_001193791.1 | 3 - 4 | 468 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |