Search
Search

rrf-1| Gene Symbol: rrf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001263626.3 | NP_001250555.1 | 16 - 17 | 3754 | 66 | |
| NM_001263627.3 | NP_001250556.1 | 15 - 16 | 3501 | 66 | ||
| GenBank mRNA | AF159144.1 | - | 16 - 17 | 3754 | 66 | |
Set Membership: |
|