Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: rnt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383147.1 | 6 - 7 | 755 | 93 | ||
GenBank mRNA | AB027412.1 | - | 6 - 7 | 759 | 93 | |
AF153275.1 | - | 6 - 7 | 695 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |