Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: eor-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029299.6 | NP_001024470.1 | 3 - 4 | 446 | 89 | |
NM_001029300.6 | NP_001024471.1 | 3 - 4 | 446 | 89 | ||
NM_001313378.4 | NP_001300307.1 | 3 | 410 | 89 | ||
GenBank mRNA | AF502570.1 | - | 3 - 4 | 393 | 89 | |
AF519109.1 | - | 3 - 4 | 371 | 89 | ||
AF519110.1 | - | 3 - 4 | 371 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |