Search
Search

clp-1| Gene Symbol: clp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379816.2 | NP_001367451.1 | 8 - 9 | 1428 | 116 | |
| NM_001379817.1 | NP_001367450.1 | 7 - 8 | 1764 | 116 | ||
| NM_066339.6 | NP_498740.2 | 7 - 8 | 1448 | 116 | ||
| NM_066340.6 | NP_498741.3 | 9 - 10 | 1661 | 116 | ||
Set Membership: |
|