Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: dys-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306310.3 | NP_001293239.1 | 16 - 17 | 3445 | 89 | |
NM_001306311.3 | NP_001293240.1 | 5 - 6 | 646 | 89 | ||
NM_001306313.4 | NP_001293242.1 | 19 - 20 | 4090 | 89 | ||
NM_001306314.3 | NP_001293243.1 | 15 - 16 | 3412 | 89 | ||
NM_001383731.1 | 15 - 16 | 3463 | 89 | |||
NM_060545.6 | NP_492946.1 | 20 - 21 | 4185 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |