Search
Search

lgc-45| Gene Symbol: lgc-45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348628.3 | NP_001335538.1 | 13 - 14 | 2484 | 65 | |
| NM_001348629.3 | NP_001335537.1 | 13 - 14 | 2444 | 65 | ||
| NM_001383812.1 | 13 - 14 | 2461 | 65 | |||
Set Membership: |
|