Search
Search

| Gene Symbol: dhs-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063099.5 | NP_495500.1 | 1 - 2 | 331 | 145 | |
| Gene Symbol: E04F6.15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063100.3 | NP_495501.1 | 4 - 5 | 1712 | 145 | |
Set Membership: |
|