Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cra-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066093.6 | NP_498494.2 | 12 - 13 | 1602 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |