Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nep-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028032.6 | NP_001023203.2 | 5 - 6 | 833 | 96 | |
NM_001028034.5 | NP_001023205.2 | 3 - 4 | 497 | 96 | ||
NM_001383127.1 | 4 - 5 | 629 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |