Search
Search

cka-1| Gene Symbol: cka-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136377.4 | NP_001129849.1 | 1 - 2 | 141 | 70 | |
| NM_001306954.3 | NP_001293883.1 | 1 - 2 | 143 | 70 | ||
| NM_069331.4 | NP_501732.1 | 2 - 3 | 219 | 70 | ||
Set Membership: |
|