Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313285.3 | NP_001300214.1 | 1 - 2 | 790 | 62 | |
NM_070750.7 | NP_503151.1 | 3 - 4 | 1082 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |