Search
Search

unc-68| Gene Symbol: unc-68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356673.4 | NP_001343696.1 | 16 | 5637 | 107 | |
| NM_001356674.3 | NP_001343811.1 | 16 | 5637 | 107 | ||
| NM_001356675.3 | NP_001343689.1 | 16 | 5637 | 107 | ||
| NM_001356676.3 | NP_001343809.1 | 16 | 5637 | 107 | ||
| NM_001356677.4 | NP_001343695.1 | 16 | 5637 | 107 | ||
| NM_001356678.4 | NP_001343810.1 | 16 | 5637 | 107 | ||
| NM_001356679.3 | NP_001343688.1 | 16 - 17 | 5726 | 107 | ||
| NM_001356680.4 | NP_001343808.1 | 16 | 5637 | 107 | ||
| NM_001356681.3 | NP_001343815.1 | 16 - 17 | 5631 | 107 | ||
| NM_001356682.3 | NP_001343819.1 | 16 - 17 | 5631 | 107 | ||
| NM_001356683.3 | NP_001343813.1 | 16 - 17 | 5631 | 107 | ||
| NM_001356684.3 | NP_001343817.1 | 16 - 17 | 5631 | 107 | ||
| NM_001356685.3 | NP_001343814.1 | 16 - 17 | 5631 | 107 | ||
| NM_001356686.3 | NP_001343818.1 | 16 - 17 | 5631 | 107 | ||
| NM_001356687.3 | NP_001343812.1 | 16 - 17 | 5631 | 107 | ||
| NM_001356688.3 | NP_001343816.1 | 16 - 17 | 5631 | 107 | ||
Set Membership: |
|