Search
Search

srx-92| Gene Symbol: srx-92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028611.3 | NP_001023782.1 | 4 - 5 | 566 | 155 | |
| NM_001028612.1 | NP_001023783.1 | 5 - 6 | 756 | 155 | ||
Set Membership: |
|