Search
Search

dkf-2| Gene Symbol: dkf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129550.2 | NP_001123022.1 | 6 - 7 | 1082 | 81 | |
| NM_001269795.3 | NP_001256724.1 | 11 - 12 | 1617 | 81 | ||
| NM_001269797.3 | NP_001256726.1 | 7 - 8 | 957 | 81 | ||
| NM_074838.6 | NP_507239.2 | 11 - 12 | 1686 | 81 | ||
Set Membership: |
|