Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: mlt-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270009.3 | NP_001256938.1 | 1 - 2 | 48 | 57 | |
NM_001270010.3 | NP_001256939.1 | 1 - 2 | 48 | 57 | ||
NM_001270011.2 | NP_001256940.1 | 1 - 2 | 71 | 57 | ||
NM_001270012.3 | NP_001256941.1 | 1 - 2 | 48 | 57 | ||
NM_001270013.3 | NP_001256942.1 | 1 - 2 | 73 | 57 | ||
NM_001270014.3 | NP_001256943.1 | 1 - 2 | 48 | 57 | ||
NM_001270015.4 | NP_001256944.1 | 1 - 2 | 77 | 57 | ||
NM_001270016.3 | NP_001256945.1 | 1 - 2 | 48 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |