Search
Search

unc-1| Gene Symbol: unc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129690.5 | NP_001123162.1 | 3 - 4 | 308 | 87 | |
| NM_075801.6 | NP_508202.1 | 3 - 4 | 290 | 87 | ||
| GenBank mRNA | EF538426.1 | - | 3 - 4 | 298 | 87 | |
Set Membership: |
|