Search
Search

irld-24| Gene Symbol: irld-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373563.3 | NP_001360674.1 | 7 - 8 | 1375 | 148 | |
| NM_077931.4 | NP_510332.1 | 7 - 8 | 1481 | 148 | ||
Set Membership: |
|