Search
Search

mps-2| Gene Symbol: mps-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038246.6 | NP_001033335.1 | 2 - 3 | 207 | 140 | |
| NM_001381312.1 | NP_001367360.1 | 3 - 4 | 450 | 140 | ||
| NM_001383777.1 | 3 - 4 | 1011 | 140 | |||
| GenBank mRNA | AY255666.1 | - | 2 - 3 | 207 | 140 | |
Set Membership: |
|