Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003347.1 | NP_001003347.1 | 413 | 70 | ||
GenBank mRNA | AB127975.1 | - | 413 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris |