Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003143.1 | NP_001003143.1 | 3 - 4 | 428 | 75 | |
XM_005618251.3 | XP_005618308.1 | 3 - 4 | 836 | 75 | ||
GenBank mRNA | AF117714.1 | - | 3 - 4 | 428 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Inventoried Probe spans exons |