Search
Search
| Gene Symbol: GAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633758.3 | XP_005633815.1 | 3 - 4 | 420 | 70 | |
| XM_022407941.1 | XP_022263649.1 | 2 - 3 | 387 | 70 | ||
| XM_534091.6 | XP_534091.2 | 3 - 4 | 326 | 70 | ||
Set Membership: |
|