Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: PLAGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615474.3 | XP_005615531.1 | 268 | 93 | ||
XM_014112545.2 | XP_013968020.1 | 268 | 93 | |||
XM_022417720.1 | XP_022273428.1 | 234 | 93 | |||
XM_022417724.1 | XP_022273432.1 | 209 | 93 | |||
XM_022417725.1 | XP_022273433.1 | 132 | 93 | |||
XM_022417744.1 | XP_022273452.1 | 268 | 93 | |||
XM_022417747.1 | XP_022273455.1 | 268 | 93 | |||
XM_022417748.1 | XP_022273456.1 | 268 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris |