Search
Search
| Gene Symbol: BOLA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014114506.2 | XP_013969981.1 | 1 - 2 | 155 | 104 | |
| XM_022419896.1 | XP_022275604.1 | 1 - 2 | 155 | 104 | ||
| XM_844690.5 | XP_849783.1 | 1 - 2 | 297 | 104 | ||
Set Membership: |
|