Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CNBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252264.1 | NP_001239193.1 | 1 | 302 | 75 | |
XM_005632022.3 | XP_005632079.1 | 1 | 298 | 75 | ||
XM_022406218.1 | XP_022261926.1 | 1 | 315 | 75 | ||
GenBank mRNA | DN371621.1 | - | 257 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |