Search
Search
| Gene Symbol: RPS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287105.1 | NP_001274034.1 | 2 - 3 | 249 | 125 | |
| NM_001287106.1 | NP_001274035.1 | 2 - 3 | 220 | 125 | ||
| GenBank mRNA | CO658560.1 | - | 3 - 4 | 361 | 125 | |
| DN863506.1 | - | 3 - 4 | 376 | 125 | ||
| DN871196.1 | - | 2 - 3 | 220 | 125 | ||
| GR889127.1 | - | 2 - 3 | 250 | 125 | ||
| GR894797.1 | - | 2 - 3 | 198 | 125 | ||
Set Membership: |
|