Search
Search

MIRLET7A-2| Gene Symbol: MIRLET7A-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049421.1 | 1 | 48 | 73 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-99a-1 | MIR99A-1 | cfa-mir-99a-1 | -566.0 | Sense | Chr. 31 - 13258782 - 13258840 on NCBI Build CanFam3.1 |
| Downstream | microRNA let-7c | MIRLET7C | cfa-let-7c | 30.0 | Sense | Chr. 31 - 13259522 - 13259588 on NCBI Build CanFam3.1 |
Set Membership: |
|