Search
Search

Ttll5| Gene Symbol: Ttll5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502772.2 | XP_003502820.1 | 11 - 12 | 1180 | 107 | |
| XM_007643400.1 | XP_007641590.1 | 11 - 12 | 1180 | 107 | ||
| XM_007643401.1 | XP_007641591.1 | 11 - 12 | 1118 | 107 | ||
| XM_007643402.1 | XP_007641592.1 | 11 - 12 | 1180 | 107 | ||
| XM_007643403.1 | XP_007641593.1 | 11 - 12 | 1180 | 107 | ||
| XM_007643404.1 | XP_007641594.1 | 11 - 12 | 1180 | 107 | ||
| XM_007643405.1 | XP_007641595.1 | 11 - 12 | 1180 | 107 | ||
| XM_007643406.1 | XP_007641596.1 | 11 - 12 | 1180 | 107 | ||
| XM_007643407.1 | XP_007641597.1 | 6 - 7 | 543 | 107 | ||
| GenBank mRNA | JI874320.1 | - | 11 - 12 | 1082 | 107 | |
| JP046762.1 | - | 11 - 12 | 1081 | 107 | ||
Set Membership: |
|