Search
Search

Hmgxb3| Gene Symbol: Hmgxb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646827.1 | XP_007645017.1 | 16 - 17 | 3061 | 66 | |
| XM_007646828.1 | XP_007645018.1 | 17 - 18 | 2965 | 66 | ||
| GenBank mRNA | JI886314.1 | - | 3 - 4 | 410 | 66 | |