Search
Search

Isg20l2| Gene Symbol: Isg20l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502903.2 | XP_003502951.1 | 1 - 2 | 1350 | 76 | |
| GenBank mRNA | JI882960.1 | - | 1 - 2 | 1323 | 76 | |
| JP060714.1 | - | 1 - 2 | 1323 | 76 | ||
Set Membership: |
|