Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Akip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509904.2 | XP_003509952.1 | 5 - 6 | 593 | 92 | |
XM_007650557.1 | XP_007648747.1 | 5 - 6 | 522 | 92 | ||
XM_007650558.1 | XP_007648748.1 | 4 - 5 | 442 | 92 | ||
GenBank mRNA | JI892296.1 | - | 5 - 6 | 577 | 92 | |
JP048937.1 | - | 5 - 6 | 577 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |