Search
Search

Dclre1b| Gene Symbol: Dclre1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505814.2 | XP_003505862.1 | 2 - 3 | 650 | 89 | |
| GenBank mRNA | JI870251.1 | - | 2 - 3 | 480 | 89 | |
| JP057033.1 | - | 2 - 3 | 480 | 89 | ||
Set Membership: |
|