Search
Search

Pde1a| Gene Symbol: Pde1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642762.1 | XP_007640952.1 | 3 - 4 | 730 | 72 | |
| XM_007642763.1 | XP_007640953.1 | 3 - 4 | 730 | 72 | ||
| XM_007642764.1 | XP_007640954.1 | 3 - 4 | 730 | 72 | ||
Set Membership: |
|