Search Thermo Fisher Scientific
Gene Symbol: Slc46a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495771.2 | XP_003495819.1 | 2 - 3 | 1158 | 80 | |
GenBank mRNA | JI878877.1 | - | 1 - 2 | 206 | 80 | |
JP060011.1 | - | 1 - 2 | 206 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |