Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grpel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496612.2 | XP_003496660.1 | 1 - 2 | 121 | 72 | |
GenBank mRNA | JI889556.1 | - | 1 - 2 | 75 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |