Search
Search

Stxbp3| Gene Symbol: Stxbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514867.2 | XP_003514915.1 | 1 - 2 | 141 | 83 | |
| XM_007654905.1 | XP_007653095.1 | 1 - 2 | 142 | 83 | ||
| GenBank mRNA | JI894043.1 | - | 1 - 2 | 134 | 83 | |
| JP062973.1 | - | 1 - 2 | 134 | 83 | ||
Set Membership: |
|