Search
Search

Gng12| Gene Symbol: Gng12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511120.2 | XP_003511168.1 | 3 - 4 | 198 | 86 | |
| XM_007651672.1 | XP_007649862.1 | 4 - 5 | 386 | 86 | ||
| XM_007651673.1 | XP_007649863.1 | 5 - 6 | 461 | 86 | ||
| GenBank mRNA | JP054680.1 | - | 2 - 3 | 176 | 86 | |
Set Membership: |
|