Search
Search

Peak1| Gene Symbol: Peak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497912.2 | XP_003497960.1 | 4 - 5 | 3734 | 69 | |
| XM_007619507.1 | XP_007617697.1 | 4 - 5 | 3645 | 69 | ||
| XM_007619517.1 | XP_007617707.1 | 4 - 5 | 3647 | 69 | ||
| XM_007619526.1 | XP_007617716.1 | 5 - 6 | 3813 | 69 | ||
| XM_007619540.1 | XP_007617730.1 | 3 - 4 | 3469 | 69 | ||
Set Membership: |
|