Search
Search

SLC35A5| Gene Symbol: SLC35A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674922.3 | XP_005674979.1 | 6 - 7 | 1464 | 74 | |
| XM_005674923.3 | XP_005674980.1 | 6 - 7 | 1458 | 74 | ||
| GenBank mRNA | JO335576.2 | - | 3 - 4 | 1025 | 74 | |
| JO419575.2 | - | 6 - 7 | 1454 | 74 | ||
| JO429276.2 | - | 6 - 7 | 1471 | 74 | ||
| JO431038.2 | - | 6 - 7 | 1471 | 74 | ||
| JO431082.2 | - | 6 - 7 | 1471 | 74 | ||
| JO501861.2 | - | 2 - 3 | 862 | 74 | ||
| JO565505.2 | - | 1 - 2 | 437 | 74 | ||
| JO586704.2 | - | 6 - 7 | 1368 | 74 | ||
| JO586706.2 | - | 6 - 7 | 1368 | 74 | ||
| JO643889.2 | - | 6 - 7 | 1474 | 74 | ||
| JO643890.2 | - | 6 - 7 | 1474 | 74 | ||
| JT717364.1 | - | 2 - 3 | 847 | 74 | ||
| KA323939.1 | - | 1 - 2 | 412 | 74 | ||
Set Membership: |
|