Search Thermo Fisher Scientific
Gene Symbol: KRTCAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046232.1 | XP_017901721.1 | 3 - 4 | 315 | 70 | |
GenBank mRNA | JO445826.2 | - | 290 | 70 | ||
JO581641.1 | - | 3 - 4 | 453 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |