Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285766.1 | NP_001272695.1 | 5 - 6 | 1018 | 98 | |
GenBank mRNA | AY510111.1 | - | 5 - 6 | 1018 | 98 | |
KA352949.1 | - | 3 - 4 | 826 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |