Search
Search

SERPINA14| Gene Symbol: SERPINA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314301.1 | NP_001301230.1 | 2 - 3 | 901 | 80 | |
| XM_005695353.3 | XP_005695410.2 | 3 - 4 | 983 | 80 | ||
| XM_013973196.2 | XP_013828650.2 | 3 - 4 | 1020 | 80 | ||
| XM_013973197.2 | XP_013828651.2 | 3 - 4 | 1271 | 80 | ||
| XM_013973198.2 | XP_013828652.2 | 3 - 4 | 939 | 80 | ||
| GenBank mRNA | AY679515.2 | - | 2 - 3 | 878 | 80 | |
| AY679517.2 | - | 2 - 3 | 365 | 80 | ||
| JO435110.2 | - | 2 - 3 | 488 | 80 | ||
Set Membership: |
|